Specifications
| Chemical Name or Material | Ruxolitinib |
| CAS | 941678-49-5 |
| Quantity | 100 mg |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C17 H18 N6 |
| Physical Form | Powder |
| Color | White to Almost-white |
| Purity | ≥97.5% |