missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RXRB Recombinant Rabbit Monoclonal Antibody (3T4L7)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558114

 View more versions of this product

Product Code. 30282953

  • 436.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MSWAARPPFL PQRHAAGQCG PVGVRKEMHC GVASRWRRRR PWLDPAAAAA AAVAGGEQQT PEPEPGEAGR DGMGDSGRDS RSPDSSSPNP LPQGVPPPSP.

This gene encodes a member of the retinoid X receptor (RXR) family of nuclear receptors which are involved in mediating the effects of retinoic acid (RA). This receptor forms homodimers with the retinoic acid, thyroid hormone, and vitamin D receptors, increasing both DNA binding and transcriptional function on their respective response elements. The gene lies within the major histocompatibility complex (MHC) class II region on chromosome 6.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RXRB
Recombinant Monoclonal
1.4 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P28702, P28704, P49743
Rxrb
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RXRB (NP_068811.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
3T4L7
Unconjugated
Rxrb
AL023085; DADB-100D22.1; DAUDI6; H-2RIIBP; MGC1831; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; NR2B2; nuclear receptor coregulator 1; nuclear receptor co-regulator 1; nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; RCoR-1; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; retinoid X receptor; retinoid X receptor beta; retinoid X receptor, beta; Rub; RXR Beta; Rxrb; RXR-beta
Rabbit
Protein A
RUO
20182, 361801, 6257
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.