missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydro-1-naphthoic Acid, TRC

Product Code. 30417341
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30417341

Brand: Toronto Research Chemicals TRCT294115500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-1,2,3,4-Tetrahydro-1-naphthoic Acid
CAS 85977-52-2
Molecular Formula C11H12O2
Quantity 500 mg
Synonym (1S)-1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid, D-(-)-1,2,3,4-Tetrahydro-1-naphthoic Acid
InChI Formula InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7H2,(H,12,13)/t10-/m0/s1
SMILES OC(=O)[C@H]1CCCc2ccccc12
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 176.21
Formula Weight 176.0837
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.