missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1,2,3,4-Tetrahydro-1-naphthoic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (S)-1,2,3,4-Tetrahydro-1-naphthoic Acid |
| CAS | 85977-52-2 |
| Molecular Formula | C11H12O2 |
| Quantity | 1 g |
| Synonym | (1S)-1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid, D-(-)-1,2,3,4-Tetrahydro-1-naphthoic Acid |
| InChI Formula | InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7H2,(H,12,13)/t10-/m0/s1 |
| SMILES | OC(=O)[C@H]1CCCc2ccccc12 |
| IUPAC Name | (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid |
| Molecular Weight (g/mol) | 176.21 |
| Formula Weight | 176.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction