missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydro-5-methoxy-N-propyl-2-naphthalenamine Hydrochloride, TRC

Product Code. 30423496
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423496 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423496 Supplier Toronto Research Chemicals Supplier No. TRCT29387525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93601-86-6
Molecular Formula C14H22ClNO
Quantity 25 mg
Synonym (S)-5-Methoxy-N-propyl-2-Aminotetralin Hydrochloride
InChI Formula InChI=1S/C14H21NO.ClH/c1-3-9-15-12-7-8-13-11(10-12)5-4-6-14(13)16-2;/h4-6,12,15H,3,7-10H2,1-2H3;1H/t12-;/m0./s1
SMILES Cl.CCCN[C@H]1CCc2c(C1)cccc2OC
IUPAC Name (2S)-5-methoxy-N-propyl-1,2,3,4-tetrahydronaphthalen-2-amine;hydrochloride
Molecular Weight (g/mol) 255.78
Formula Weight 255.139
Chemical Name or Material (S)-1,2,3,4-Tetrahydro-5-methoxy-N-propyl-2-naphthalenamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.