missing translation for 'onlineSavingsMsg'
Learn More

S-(1-Methyl-2-hydroxyethyl)glutathione(Mixture of Diastereomers), TRC

Product Code. 30465106
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465106 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465106 Supplier Toronto Research Chemicals Supplier No. TRCM3126351MG
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material S-(1-Methyl-2-hydroxyethyl)glutathione(Mixture of Diastereomers)
CAS 1356019-76-5
Molecular Formula C13H23N3O7S
Quantity 1 mg
Synonym L-γ-Glutamyl-S-(1-methyl-2-hydroxyethyl)-L-cysteinylgycine, N-[N-L-γ-glutamyl-S-(1-methyl-2-hydroxyethyl)-L-cysteinyl]glycine
InChI Formula InChI=1S/C13H23N3O7S/c1-7(5-17)24-6-9(12(21)15-4-11(19)20)16-10(18)3-2-8(14)13(22)23/h7-9,17H,2-6,14H2,1H3,(H,15,21)(H,16,18)(H,19,20)(H,22,23)/t7?,8-,9-/m0/s1
SMILES OCC(C)SC[C@@H](C(NCC(O)=O)=O)NC(CC[C@@H](C(O)=O)N)=O
IUPAC Name N5-((2R)-1-((carboxymethyl)amino)-3-((1-hydroxypropan-2-yl)thio)-1-oxopropan-2-yl)-L-glutamine
Molecular Weight (g/mol) 365.4
Formula Weight 365.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.