missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Trichloromethyl-1,2,3,4-Tetrahydro-Beta-carboline Hydrochloride, TRC

Product Code. 30457105
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457105 1 mg 1mg
30456702 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457105 Supplier Toronto Research Chemicals Supplier No. TRCT7742401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773057-50-4
Molecular Formula C12H11Cl3N2 . HCl
Molecular Weight (g/mol) 326
Synonym (S)-2,3,4,9-Tetrahydro-1-(trichloromethyl)-1H-pyrido[3,4-b]indole Hydrochloride, (R)-TaClo Hydrochloride
IUPAC Name (1S)-1-(trichloromethyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole;hydrochloride
SMILES Cl.ClC(Cl)(Cl)[C@H]1NCCc2c1[nH]c3ccccc23

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H11Cl3N2.ClH/c13-12(14,15)11-10-8(5-6-16-11)7-3-1-2-4-9(7)17-10;/h1-4,11,16-17H,5-6H2;1H/t11-;/m0./s1
Formula Weight 323.9755
Chemical Name or Material (S)-1-Trichloromethyl-1,2,3,4-Tetrahydro-beta-carboline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.