missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Aminobutyramide Hydrochloride, TRC

Product Code. 17933985
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17933985 1 g 1g
30133086 25 g 25g
30137219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933985 Supplier Toronto Research Chemicals Supplier No. TRCA6028901G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7682-20-4
Molecular Formula C4 H10 N2 O . Cl H
Molecular Weight (g/mol) 138.6
Synonym Butanamide, 2-amino-, hydrochloride (1:1), (2S)-, Butanamide, 2-amino-, monohydrochloride, (2S)- (9CI), Butanamide, 2-amino-, monohydrochloride, (S)-, Butyramide, 2-amino-, monohydrochloride, L- (8CI), (S)-(+)-2-Aminobutanamide hydrochloride, (S)-2-Aminobutanamide hydrochloride, (S)-2-Aminobutyramide hydrochloride, Levetiracetam Related Compound B, Levetiracetam Imp. G (EP)
IUPAC Name (2S)-2-aminobutanamide;hydrochloride
SMILES Cl.CC[C@H](N)C(=O)N

Spécification

InChI Formula InChI=1S/C4H10N2O.ClH/c1-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H2,6,7);1H/t3-;/m0./s1
Quantity 1 g
Formula Weight 138.056
Shipping Condition Room Temperature
Chemical Name or Material (S)-2-Aminobutanamide Hydrochloride
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.