missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylbutyric acid, 99%

Product Code. 11323356
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323356 1 g 1g
11333356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323356 Supplier Thermo Scientific Alfa Aesar Supplier No. L12754.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4286-15-1 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4286-15-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00063164
InChI Key OFJWFSNDPCAWDK-VIFPVBQESA-N
Synonym s-+-2-phenylbutyric acid, s-2-phenylbutanoic acid, 2s-2-phenylbutanoic acid, s-2-phenylbutyric acid, pubchem9874, 2-phenylbutyric acid, s, s--2-phenyl-mutyric acid, s-+-2-phenylbutanoic acid, s-+-2-phenyl-mutyric acid
PubChem CID 785329
IUPAC Name (2S)-2-phenylbutanoic acid
SMILES CCC(C1=CC=CC=C1)C(=O)O

Specifications

Density 1.055
Boiling Point 270°C to 272°C
Flash Point >110°C (230°F)
Refractive Index 1.515
Quantity 1 g
Beilstein 2045208
Optical Rotation +100° (c=5 in Toluene)
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Phenylbutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.