missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid, 97%, BLD Pharm™

Product Code. 31033884
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31033884 1 g -
31021135 100 mg -
31023293 250 mg -
31029781 5 g 5g
31025325 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31033884 Supplier BLD Pharm Supplier No. BD123991g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 102507-13-1
Molecular Formula C10H19NO5
MDL Number MFCD03094792
Quantity 1 g
SMILES CC(C)(O)[C@H](NC(OC(C)(C)C)=O)C(O)=O
Molecular Weight (g/mol) 233.26
Percent Purity 97%
Chemical Name or Material (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.