missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%, BLD Pharm™

Product Code. 31030759
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
250mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31030759 1 g -
31020978 25 g -
31021941 10 g -
31021148 5 g -
31014924 100 g 100g
31025414 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31030759 Supplier BLD Pharm Supplier No. BD007991251g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 102308-32-7
Molecular Formula C11H19NO4
MDL Number MFCD00209557
Quantity 1 g
SMILES O=C(N1C(C)(C)OC[C@H]1C=O)OC(C)(C)C
Molecular Weight (g/mol) 229.27
Percent Purity 95%
Chemical Name or Material (S)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.