missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 95%

Product Code. 15429585
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429585 250 mg 250mg
15409585 1 g 1g
15419585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15429585 Supplier Thermo Scientific Alfa Aesar Supplier No. H52101.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 507472-25-5 | C24H20N2O6 | 432.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507472-25-5
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD03428007
InChI Key DRESTDPDCGPKNI-UHFFFAOYNA-N
Synonym s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-s-3-amino-3-2-nitro-phenyl-propionic acid, fmoc-s-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24027, fmoc-?-phe 2-no2-oh, n-fmoc-2-nitro-d-beta-phenylalanine, fmoc-s-3-amino-3-2-nitrophenyl propionic acid
PubChem CID 7023399
IUPAC Name 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
SMILES OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 154°C to 156°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 432.43
Percent Purity 95%
Chemical Name or Material (S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.