Learn More
(S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione (7-Ethyl-20(S)-camptothecin), Mikromol™
Code produit 30052086
Modifier l'affichage
Click to view available options
Quantity:
25 mg
Conditionnement:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
30052086 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30052086 Fournisseur Mikromol Code fournisseur MM1157.050025
 Restricted Item
Estimated Shipment: 30-11-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione (7-Ethyl-20(S)-camptothecin)
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material 7-Ethyl-20(S)-camptothecin
CAS 78287-27-1
Analyte or Component Names 7-Ethyl-20(S)-camptothecin
Shipping Condition Room Temperature
Molecular Formula C22 H20 N2 O4
Certifications/Compliance ISO 17025
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4,11-diethyl-4-hydroxy-, (4S)-, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4,11-diethyl-4-hydroxy-, (S)-, 7-Ethyl-20(S)-camptothecin, 7-Ethylcamptothecin, SN 22, Irinotecan Related Compound E, Irinotecan Hydrochloride Trihydrate Impurity F, Irinotecan Impurity F
InChI Formula InChI=1S/C22H20N2O4/c1-3-12-13-7-5-6-8-17(13)23-19-14(12)10-24-18(19)9-16-15(20(24)25)11-28-21(26)22(16,27)4-2/h5-9,27H,3-4,10-11H2,1-2H3/t22-/m0/s1
SMILES CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccccc15)[C@@](O)(CC)C(=O)OC4
IUPAC Name (4S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
Molecular Weight (g/mol) 376.41
Quantity 25 mg
Formula Weight 376.1423 g/mol