missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one, TRC

Product Code. 30136810
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136810 100 mg 100mg
30132688 10 mg 10mg
30010803 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136810 Supplier Toronto Research Chemicals Supplier No. TRCA576955100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446292-10-0
Molecular Formula C14 H17 N3 O4
Molecular Weight (g/mol) 291.3
Synonym 3-Morpholinone, 4-[4-[(5S)-5-(aminomethyl)-2-oxo-3-oxazolidinyl]phenyl]- (9CI, ACI), 4-[4-[(5S)-5-(Aminomethyl)-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone (ACI), (S)-4-[4-(5-Aminomethyl-2-oxooxazolidin-3-yl)phenyl]morpholin-3-one, 4-[4-((S)-5-[Aminomethyl]-2-oxooxazolidin-3-yl)phenyl]morpholine-3-one, 4-[4-[(5S)-5-(Aminomethyl)-2-oxo-1,3-oxazolidin-3-yl]phenyl]-morpholin-3-one, 4-[4-[(5S)-5-(Aminomethyl)-2-oxo-1,3-oxazolidin-3-yl]phenyl]morpholin-3-one
IUPAC Name 4-[4-[(5S)-5-(aminomethyl)-2-oxo-1,3-oxazolidin-3-yl]phenyl]morpholin-3-one
SMILES C1=C(C=CC(=C1)N2C[C@H](CN)OC2=O)N3CCOCC3=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H17N3O4/c15-7-12-8-17(14(19)21-12)11-3-1-10(2-4-11)16-5-6-20-9-13(16)18/h1-4,12H,5-9,15H2/t12-/m0/s1
Formula Weight 291.1219
Chemical Name or Material 4-[4-[(5S)-5-(Aminomethyl)-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone (Rivaroxaban)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.