Learn More
(S)-4-Benzyl-3-propionyl-2-oxazolidinone, TRC
Product Code. 30335926
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30335926 Supplier Toronto Research Chemicals Supplier No. TRCB2886501G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-4-Benzyl-3-propionyl-2-oxazolidinone
CAS 101711-78-8
Molecular Formula C13H15NO3
Quantity 1 g
Synonym (4S)-3-(1-Oxopropyl)-4-(phenylmethyl)-2-oxazolidinone, (+)-4-Benzyl-3-propionyl-2-oxazolidinone, (4S)-4-Benzyl-3-propanoyloxazolidin-2-one, (4S)-4-Benzyl-3-propionyl-1,3-oxazolidin-2-one, (S)-N-Propionyl-4-benzyl-1,3-oxazolidin-2-one, 3-[1-Oxopropyl]-4(S)-(phenylmethyl)-2-oxazolidinone
InChI Formula InChI=1S/C13H15NO3/c1-2-12(15)14-11(9-17-13(14)16)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3/t11-/m0/s1
SMILES CCC(=O)N1[C@@H](Cc2ccccc2)COC1=O
IUPAC Name (4S)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 233.26
Formula Weight 233.1052