missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-6-Methyl-5-hepten-2-ol, 99%

Product Code. 11309545
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309545 250 mg 250mg
11399535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309545 Supplier Thermo Scientific Alfa Aesar Supplier No. L18884.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58917-26-3 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58917-26-3
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD03093080
InChI Key OHEFFKYYKJVVOX-QMMMGPOBSA-N
Synonym s-sulcatol, s-6-methylhept-5-en-2-ol, 2s-6-methylhept-5-en-2-ol, 2s-6-methyl-5-hepten-2-ol, s-+-6-methyl-5-hepten-2-ol, 6-methyl-5-hepten-2s-ol, 5-hepten-2-ol, 6-methyl-, 2s, +-sulcatol, s-6-methyl-hept-5-en-2-ol
PubChem CID 6971127
IUPAC Name (2S)-6-methylhept-5-en-2-ol
SMILES CC(CCC=C(C)C)O

Specifications

Density 0.844
Boiling Point 78°C to 80°C (15 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.448
Quantity 250 mg
Beilstein 1720071
Optical Rotation +18° (Neat)
Formula Weight 128.22
Percent Purity 99%
Chemical Name or Material (S)-(+)-6-Methyl-5-hepten-2-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.