Learn More
Mecmesin™ S-Beam Load Cell
Product Code. 11913858
Encompass
Change view
Click to view available options
Capacity (Metric):
10,000 N
500 N
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11913858 500 N Each
11968019 10,000 N Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11913858 Supplier Mecmesin™ Supplier No. 870009
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
R\u00E9cupel-bijdrage uitgesloten
Mecmesin's S-Beam Loadcell is a flexible solution for general force regulation applications.
  • This is suitable for areas where space is not limited.
  • The unit features a peak hold of maximum reading at a fast rate of 5000 Hz when used with AFTI display and advanced force gauge.
  • The unit is also applicable for use in tension and compression testing.
Specifications
Capacity (Metric) 500 N