missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Benzyl-DL-cysteine-2,3,3-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | S-Benzyl-DL-cysteine-2,3,3-D3 |
| CAS | 51494-04-3 |
| Molecular Formula | C10 D3 H10 N O2 S |
| Quantity | 0.1 g |
| Synonym | Cysteine-2,3,3-d3, S-(phenylmethyl)- (9 CI), DL-Cysteine-2,3,3-d3, S-(phenylmethyl)- (ZCI), S-(Phenylmethyl)-DL-cysteine-2,3,3-d3, S-Benzyl-DL-cysteine-2,3,3-d3, S-Benzyl-DL-cysteine-2,3,3-D3 |
| InChI Formula | InChI=1 S/C10H13NO2S/c11-9(10(12)13)7-14-6-8-4-2-1-3-5-8/h1-5,9 H,6-7,11H2,(H,12,13)/i7D2,9 D |
| SMILES | [2 H]C([2 H])(SCc1ccccc1)C([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-3-benzylsulfanyl-2,3,3-trideuteriopropanoic acid |
| Molecular Weight (g/mol) | 214.299 |
| Formula Weight | 214.086 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction