Learn More
(S)-Citalopram Oxalate, TRC
Product Code. 30135706
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135706 1 g 1g
17970975 100 mg 100mg
17924155 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135706 Supplier Toronto Research Chemicals Supplier No. TRCC5050101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 219861-08-2
Molecular Formula C20 H21 F N2 O . C2 H2 O4
Molecular Weight (g/mol) 414.43
Synonym Escitalopram Oxalate, (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile ethanedioate, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, (1S)-, ethanedioate (1:1), (+)-(S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile oxalate (1:1), (S)-(+)Citalopram oxalate, Cipra-Pro, Cipralex, (S)-Citalopram oxalate, Lexapro, Lu 26-054-0, Szetalo
IUPAC Name (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
SMILES CN(C)CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O
Spécification
Quantity 1 g
InChI Formula InChI=1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m0./s1
Formula Weight 414.1591
Percent Purity >95%
Chemical Name or Material Escitalopram Oxalate
Grade HPLC