missing translation for 'onlineSavingsMsg'
Learn More

S-Desethyl S-Methyl Amisulpride, TRC

Product Code. 17991055
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991055 5 mg 5mg
17911625 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991055 Supplier Toronto Research Chemicals Supplier No. TRCD2895955MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71676-00-1
Molecular Formula C16 H25 N3 O4 S
Molecular Weight (g/mol) 355.45
Synonym 4-Amino-N-[[(2RS)-1-ethyl-pyrrolidin-2-yl]methyl]-2-methoxy-5-(methylsulphonyl)benzamide, Amisulpride Imp. D (EP)
IUPAC Name 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-methylsulfonylbenzamide
SMILES CCN1CCCC1CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H25N3O4S/c1-4-19-7-5-6-11(19)10-18-16(20)12-8-15(24(3,21)22)13(17)9-14(12)23-2/h8-9,11H,4-7,10,17H2,1-3H3,(H,18,20)
Formula Weight 355.1566
Chemical Name or Material 4-Amino-N-[[(2RS)-1-ethyl-pyrrolidin-2-yl]methyl]-2-methoxy-5-(methylsulphonyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.