Learn More
(S)-Methocarbamol, TRC
Product Code. 30474962
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474962 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30474962 Supplier Toronto Research Chemicals Supplier No. TRCM225980250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-Methocarbamol
CAS 108914-10-9
Molecular Formula C11 H15 N O5
Quantity 250 mg
Synonym 1,2-Propanediol, 3-(2-methoxyphenoxy)-, 1-carbamate, (2S)- (9CI, ACI), 1,2-Propanediol, 3-(2-methoxyphenoxy)-, 1-carbamate, (S)- (ZCI), Carbamic acid, (-)-2-hydroxy-3-(o-methoxyphenoxy)propyl ester (7CI), (S)-Methocarbamol, (2S)-3-(2-Methoxyphenoxy)-1,2-propanediol 1-carbamate, (S)-3-(2-Methoxyphenoxy)-1-glyceryl carbamate
InChI Formula InChI=1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)/t8-/m0/s1
SMILES COc1ccccc1OC[C@H](O)COC(=O)N
IUPAC Name [(2S)-2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate
Molecular Weight (g/mol) 241.24
Formula Weight 241.095