missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl-isothiouronium Hemisulfate, TRC

Product Code. 30484123
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30484123 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30484123 Supplier Toronto Research Chemicals Supplier No. TRCM31496010MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material S-Methyl-isothiouronium Hemisulfate
CAS 867-44-7
Molecular Formula C2H6N2S.1/2H2SO4
Quantity 10 mg
Synonym Carbamimidothioic Acid Methyl Ester Sulfate, S-Methyl-isothiouronium Sulfate
InChI Formula InChI=1S/2C2H6N2S.H2O4S/c2*1-5-2(3)4;1-5(2,3)4/h2*1H3,(H3,3,4);(H2,1,2,3,4)
SMILES CSC(=N)N.CSC(=N)N.OS(=O)(=O)O
IUPAC Name methyl carbamimidothioate;sulfuric acid
Molecular Weight (g/mol) 139.14
Formula Weight 278.0177
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.