missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl N,N-Diethylthiocarbamate, TRC

Product Code. 30137679
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137679 50 mg 50mg
30133593 100 mg 100mg
17923225 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137679 Supplier Toronto Research Chemicals Supplier No. TRCM30173050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37174-63-3
Molecular Formula C6 H13 N O S
Molecular Weight (g/mol) 147.24
Synonym Disulfiram Impurity 3
IUPAC Name S-methyl N,N-diethylcarbamothioate
SMILES CCN(CC)C(=O)SC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H13NOS/c1-4-7(5-2)6(8)9-3/h4-5H2,1-3H3
Formula Weight 147.0718
Percent Purity >95%
Chemical Name or Material S-Methyl-N,N-diethylthiocarbamate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.