missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-N-3-Benzylnirvanol, TRC

Product Code. 17984485
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984485 10 mg 10mg
17941725 50 mg 50mg
17980365 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984485 Supplier Toronto Research Chemicals Supplier No. TRCB28577510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 790676-40-3
Molecular Formula C18 H18 N2 O2
Molecular Weight (g/mol) 294.35
IUPAC Name (5S)-3-benzyl-5-ethyl-5-phenylimidazolidine-2,4-dione
SMILES CC[C@]1(NC(=O)N(Cc2ccccc2)C1=O)c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H18N2O2/c1-2-18(15-11-7-4-8-12-15)16(21)20(17(22)19-18)13-14-9-5-3-6-10-14/h3-12H,2,13H2,1H3,(H,19,22)/t18-/m0/s1
Formula Weight 294.1368
Percent Purity >95%
Chemical Name or Material (S)-(+)-N-3-Benzylnirvanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.