missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-alpha-Allylglycine dicyclohexylamine salt, 98%, 98% ee, Thermo Scientific™

Product Code. 11975521 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11975521

Brand: Thermo Scientific Acros 441160010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 143979-15-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 123°C to 124°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C12H23N·C10H17NO4
Quantity 1 g
Specific Rotation 26
Synonym dicyclohexylamine s-2-tert-butoxycarbonyl amino pent-4-enoate, boc-l-2-allylglycine dicyclohexylamine salt, boc-l-allylglycine.dcha, boc-l-allylglycine-dcha, boc-l-allylglycine dicyclohexylaminoium salt, boc-d-allylglycine-dcha, 2s-2-tert-butoxycarbonyl amino pent-4-enoic acid; dicha, boc-l-allylglycine dcha, boc-algly-oh dcha, boc-l-allyglycine?cha
InChI Key VMCGMPITVQIMGK-ZLTKDMPESA-N
SMILES CC(C)(C)OC(=O)NC(CC=C)C(=O)O.C1CCC(CC1)NC2CCCCC2
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
Molecular Weight (g/mol) 396.58
PubChem CID 2755982
Formula Weight 396.58
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-N-BOC-α-Allylglycine dicyclohexylamine salt, 98% ee
Show More Show Less
Product Identifier
  • (R)-alpha-Allylalanine hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.