missing translation for 'onlineSavingsMsg'
Learn More

(S)-Phenylephrine Hydrochloride, TRC

Product Code. 30003737
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003737 100 mg 100mg
30004269 250 mg 250mg
30005340 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003737 Supplier Toronto Research Chemicals Supplier No. TRCP320630100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-38-8
Molecular Formula C9 H13 N O2 . Cl H
Molecular Weight (g/mol) 203.67
Synonym Benzenemethanol, 3-hydroxy-α-[(methylamino)methyl]-, hydrochloride (1:1), (αS)-, Benzenemethanol, 3-hydroxy-α-[(methylamino)methyl]-, hydrochloride, (S)-, Benzenemethanol, 3-hydroxy-α-[(methylamino)methyl]-, hydrochloride, (αS)- (9CI), Benzyl alcohol, m-hydroxy-α-[(methylamino)methyl]-, hydrochloride, (+)- (8CI), (+)-Phenylephrine hydrochloride, (S)-Phenylephrine hydrochloride, (1S)-1-(3-Hydroxyphenyl)-2-(N-methylamino)ethanol hydrochloride, N-[(2S)-2-Hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylamine hydrochloride, (+)-m-Hydroxy-α-[(methylamino)methyl]benzyl alcohol hydrochloride, D-Phenylephedrine hydrochloride, (αS)-3-Hydroxy-α-[(methylamino)methyl]benzenemethanol Hydrochloride (1:1)
IUPAC Name 3-[(1S)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
SMILES Cl.CNC[C@@H](O)c1cccc(O)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/t9-;/m1./s1
Formula Weight 203.0713
Percent Purity >95%
Chemical Name or Material (S)-Phenylephrine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.