Learn More
S-Phos, TRC
Specifications
Specifications
| Chemical Name or Material | S-Phos |
| CAS | 657408-07-6 |
| Molecular Formula | C26H35O2P |
| Quantity | 1 g |
| Synonym | (2,6-Dimethoxy-1,1'-biphenyl-2-yl)dicyclohexylphosphine, 2-(Dicyclohexylphosphino)-2',6'-dimethoxy-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, Dicyclohexyl(2',6'-dimethoxy-1,1'-biphenyl-2-yl)phosphine, Dicyclohexyl(2',6'-dimethoxybiphenyl-2-yl)phosphine |
| InChI Formula | InChI=1S/C26H35O2P/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21/h9-11,16-21H,3-8,12-15H2,1-2H3 |
| SMILES | COc1cccc(OC)c1c2ccccc2P(C3CCCCC3)C4CCCCC4 |
| IUPAC Name | dicyclohexyl-[2-(2,6-dimethoxyphenyl)phenyl]phosphane |
| Molecular Weight (g/mol) | 410.53 |
| Formula Weight | 410.2375 |