missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Thalidomide, TRC

Product Code. 30451435
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451435 10 mg 10mg
30449987 25 mg -
30451877 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451435 Supplier Toronto Research Chemicals Supplier No. TRCT33886010MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 841-67-8
Molecular Formula C13 H10 N2 O4
Molecular Weight (g/mol) 258.23
Synonym 1H-Isoindole-1,3(2H)-dione, 2-[(3S)-2,6-dioxo-3-piperidinyl]- (9CI, ACI), 1H-Isoindole-1,3(2H)-dione, 2-(2,6-dioxo-3-piperidinyl)-, (S)- (ZCI), 2-[(3S)-2,6-Dioxo-3-piperidinyl]-1H-isoindole-1,3(2H)-dione (ACI), Phthalimide, N-(2,6-dioxo-3-piperidyl)-, L-(-)- (8CI), (-)-Thalidomide, (S)-(-)-Thalidomide, (S)-Thalidomide, 2-[(3S)-2,6-Dioxopiperidin-3-yl]-2,3-dihydro-1H-isoindole-1,3-dione, NSC 91730, S-(L)-Thalidomide, S-Lenalidomide
IUPAC Name 2-[(3S)-2,6-dioxopiperidin-3-yl]isoindole-1,3-dione
SMILES O=C1CC[C@H](N2C(=O)c3ccccc3C2=O)C(=O)N1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)/t9-/m0/s1
Formula Weight 258.064
Chemical Name or Material (S)-(-)-Thalidomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.