Learn More
S-(-)-Warfarin-d5, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-Warfarin phenyl-d5 |
| CAS | 791013-22-4 |
| Molecular Formula | C19 2H5 H11 O4 |
| Quantity | 1 mg |
| Synonym | 2H-1-Benzopyran-2-one','4-hydroxy-3-[3-oxo-1-(phenyl-d5)butyl]-','(S)- (9CI), (S)-4-Hydroxy-3-[3-oxo-1-(phenyl-d5)butyl]-2H-1-benzopyran-2-one, (S)-Warfarin D5 (phenyl-D5) |
| InChI Formula | InChI=1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3/t15-/m0/s1/i2D,3D,4D,7D,8D |
| SMILES | [2H]c1c([2H])c([2H])c([C@H](CC(=O)C)C2=C(O)c3ccccc3OC2=O)c([2H])c1[2H] |
| IUPAC Name | 4-hydroxy-3-[(1S)-3-oxo-1-(2,3,4,5,6-pentadeuteriophenyl)butyl]chromen-2-one |
| Molecular Weight (g/mol) | 313.36 |
| Formula Weight | 313.1362 |