Learn More
[S-(Z)]-[2-[5-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-2-methylenecyclohexylidene]ethyl]diphenylphosphine Oxide, TRC
Product Code. 30372211
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372211 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30372211 Supplier Toronto Research Chemicals Supplier No. TRCD446780100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S-(Z))-(2-(5-(((1,1-Dimethylethyl)dimethylsilyl)oxy)-2-methylenecyclohexylidene)ethyl)diphenylphosphine Oxide
CAS 100858-27-3
Molecular Formula C27H37O2PSi
Quantity 100 mg
Synonym [(2Z)-2-[(5S)-5-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-2-methylenecyclohexylidene]ethyl]diphenylphosphine Oxide
InChI Formula InChI=1S/C27H37O2PSi/c1-22-17-18-24(29-31(5,6)27(2,3)4)21-23(22)19-20-30(28,25-13-9-7-10-14-25)26-15-11-8-12-16-26/h7-16,19,24H,1,17-18,20-21H2,2-6H3/b23-19-/t24-/m0/s1
SMILES CC(C)(C)[Si](C)(C)O[C@H]1CCC(=C)\C(=C/CP(=O)(c2ccccc2)c3ccccc3)\C1
IUPAC Name tert-butyl-[(1S,3Z)-3-(2-diphenylphosphorylethylidene)-4-methylidenecyclohexyl]oxy-dimethylsilane
Molecular Weight (g/mol) 452.64
Formula Weight 452.23