Learn More
[S-(Z)]-[2-[5-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-2-methylenecyclohexylidene]ethyl]diphenylphosphine Oxide, TRC
Product Code. 30376206
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376206 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30376206 Supplier Toronto Research Chemicals Supplier No. TRCD44678010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S-(Z))-(2-(5-(((1,1-Dimethylethyl)dimethylsilyl)oxy)-2-methylenecyclohexylidene)ethyl)diphenylphosphine Oxide
CAS 100858-27-3
Molecular Formula C27H37O2PSi
Quantity 10 mg
Synonym [(2Z)-2-[(5S)-5-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-2-methylenecyclohexylidene]ethyl]diphenylphosphine Oxide
InChI Formula InChI=1S/C27H37O2PSi/c1-22-17-18-24(29-31(5,6)27(2,3)4)21-23(22)19-20-30(28,25-13-9-7-10-14-25)26-15-11-8-12-16-26/h7-16,19,24H,1,17-18,20-21H2,2-6H3/b23-19-/t24-/m0/s1
SMILES CC(C)(C)[Si](C)(C)O[C@H]1CCC(=C)\C(=C/CP(=O)(c2ccccc2)c3ccccc3)\C1
IUPAC Name tert-butyl-[(1S,3Z)-3-(2-diphenylphosphorylethylidene)-4-methylidenecyclohexyl]oxy-dimethylsilane
Molecular Weight (g/mol) 452.64
Formula Weight 452.23