Learn More
S100 beta (S100B) Monoclonal Antibody (OTI1A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF814504
Description
S100 beta (S100B) Monoclonal antibody specifically detects S100 beta (S100B) in Virus samples. It is validated for ELISASpecifications
| S100 beta (S100B) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Lyophilized |
| ELISA | |
| OTI1A3 | |
| Unconjugated | |
| AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P | |
| Recombinant fusion protein of SARS-CoV-2 Spike RBD protein and virion surface domain of membrane protein (M-protein) produced in HEK293 | |
| 100 μg | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.