missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A10 Antibody (344015) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2377JF635
This item is not returnable.
View return policy
Description
S100A10 Monoclonal antibody specifically detects S100A10 in Mouse samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| S100A10 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)) | |
| E. coli-derived recombinant mouse S100A10, Met1-Lys97, Accession # P08207 | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 6281 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 344015 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction