missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A13 Monoclonal Antibody (854114)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561421
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein is widely expressed in various types of tissues with a high expression level in thyroid gland. In smooth muscle cells, this protein co-expresses with other family members in the nucleus and in stress fibers, suggesting diverse functions in signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| S100A13 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q99584 | |
| S100a13 | |
| E. coli-derived recombinant human S100A13, Ala2-Lys98. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 854114 | |
| Unconjugated | |
| S100a13 | |
| Protein S100-A13; S100 calcium binding protein A13; S100 calcium-binding protein A13; S100A1; S100a13 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6284 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction