missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12343-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown.Specifications
| S100A3 | |
| Polyclonal | |
| Unconjugated | |
| S100A3 | |
| Protein S 100E, Protein S100 A3, S100A3, S100E | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20197, 6274 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33764, P62818 | |
| S100A3 | |
| S100A3 Fusion Protein Ag2995 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction