missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio S100A4 Mouse, Clone: S100A4/1481, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6275-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
S100A4 | |
Cytoplasmic and Nuclear | |
S100A4/1481 | |
Unconjugated | |
NP_062427 | |
Mouse | |
10-12kDa | |
Protein A/G Purified | |
RUO | |
6275 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
S100A4 | |
Recombinant human S100A4 protein | |
HeLa, A549 or A375 Cells. Placenta | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |