missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A7/Psoriasin Antibody (47C1068) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 47 publications
Brand: Novus Biologicals NB100-56559
This item is not returnable.
View return policy
Description
S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Flow (Intracellular).
Specifications
| S100A7/Psoriasin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Simple Western 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 26225121), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy reported in scientific literature (PMID 26225121), Flow (Intracellular) | |
| HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6278 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Flow Cytometry | |
| 47C1068 | |
| Unconjugated | |
| P31151 | |
| S100A7 | |
| This antibody was developed against recombinant psoriasin/HID-5 protein. | |
| 0.1 mg | |
| Primary | |
| In confluent MCF10A cells, an 11 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction