missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Mouse anti-Bovine, Human, Mouse, Rat, Clone-SPM354, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54399-100UG
This item is not returnable.
View return policy
Description
S100B Monoclonal specifically detects S100B in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| S100B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
| Mouse | |
| 11 kDa | |
| 100 μg | |
| Apoptosis, Biologically Active Proteins, Cytoskeleton Markers, Neuroscience, Stem Cells | |
| 6285 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| SPM354 | |
| Unconjugated | |
| PBS with No Preservative | |
| S100B | |
| Purified bovine brain S100 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction