missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SA 4503 Dihydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 1-[2-(3,4-Dimethoxyphenyl)ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride |
| CAS | 165377-44-6 |
| Molecular Formula | C23H32N2O2 . HCl |
| Quantity | 10 mg |
| Synonym | 1-[2-(3,4-dimethoxyphenyl)ethyl]-4-(3-phenylpropyl)-piperazine Hydrochloride (1:2), SA 4503, Cutamesine Dihydrochloride, SA 4503 Dihydrochloride |
| InChI Formula | InChI=1S/C23H32N2O2.2ClH/c1-26-22-11-10-21(19-23(22)27-2)12-14-25-17-15-24(16-18-25)13-6-9-20-7-4-3-5-8-20;;/h3-5,7-8,10-11,19H,6,9,12-18H2,1-2H3;2*1H |
| SMILES | Cl.Cl.COc1ccc(CCN2CCN(CCCc3ccccc3)CC2)cc1OC |
| IUPAC Name | 1-[2-(3,4-dimethoxyphenyl)ethyl]-4-(3-phenylpropyl)piperazine;dihydrochloride |
| Molecular Weight (g/mol) | 441.4 |
| Formula Weight | 440.1997 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction