Learn More
Ansell Edmont™ Safe-Knit™ GP 72-165 Series Gray Polyester/Glass Fiber Heavy Weight Gloves
Shop All Ansell ProductsSpecifications
Specifications
| Back Material | Polyester/Glass Fiber |
| Construction | Seamless Knit |
| Cuff Material | Polyester/Glass Fiber |
| Product Type | Heavyweight Gloves |
| Size | 10 |
| Style | 72-165 |
| Glove Cuff Color | Gray |
| Glove Cuff Style | Attached Sleeve |
| Lining | Dyneema™ |
| Quantity | 6 Pairs |
| Show More |