Description
- Accessory for Safety Waste Cap 'LISA,' V3.0
- Extension Funnel with Hinged Lid
- Made of PTFE for optimal chemical resistance
Specifications
Specifications
| For Use With (Equipment) | HPLC / UHPLC Systems |
| Includes | Extension Funnel with Earthing Cable |
| Material | PTFE |
| Thread Size | M30x35 |
| Type | Electrically Conductive |