Learn More
MEDICOM Safewear CPE Splash Gown
Product Code. 31781314 Shop All Medicom Products
Change view
Click to view available options
Size:
Universal
Unit Size:
Pack of 75
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781314 Universal Pack of 75
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781314 Supplier MEDICOM Supplier No. J50656
  • ONLINE EXCLUSIVE PRICE
    60.07€
    49.24€ / Pack of 75
    Save 10.83€
    18% Off
Estimated Shipment: 11-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The CPE hygiene gown provides superior protection to a traditional apron by covering the body and arms with secure fastening.
  • Offers superior protection compared to a traditional apron
  • Covers almost the entire body including the arms
  • Attaches with stretchable ties
  • Thumb pass system to prevent sleeves from riding up
  • Adapts to different body shapes
Specifications
Size Universal
Product Type Isolation Gown
Material CPE
Color Blue
Closure Type Back Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic