Learn More
MEDICOM Safewear PP Collared Coveralls, Zip
Product Code. 31781323 Shop All Medicom Products
Change view
Click to view available options
Size:
4X-Large
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781323 4X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781323 Supplier MEDICOM Supplier No. 40.101/07
  • ONLINE EXCLUSIVE PRICE
    44.63€
    36.59€ / Pack of 25
    Save 8.04€
    18% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SAFEWEAR polypropylene coverall is lightweight and breathable, with collar and zip closure, ideal for hygiene and HACCP programs.
  • Single-use hygiene combinations protect the work environment from particles coming from the operator's body (scales, hair, sweat, etc.)
  • They are suitable for HACCP approaches,
  • They allow the operator to protect themselves from the risks of soiling related to the activity they perform
Specifications
Product Type Single-use Coveralls
Size 4X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Welded
Sterility Non-sterile
Resistance None