missing translation for 'onlineSavingsMsg'
Learn More

Salannin, TRC

Product Code. 30136999
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136999 5 mg 5mg
30004962 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136999 Supplier Toronto Research Chemicals Supplier No. TRCS0852455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-20-1
Molecular Formula C34 H44 O9
Molecular Weight (g/mol) 596.71
Synonym Azadriactin, Margosan-O
SMILES COC(=O)C[C@@H]1[C@@]2(C)[C@H](C[C@@H](OC(=O)C)[C@@]3(C)CO[C@@H]([C@H]4O[C@@H]5C[C@H](C(=C5[C@@]14C)C)c6cocc6)[C@H]23)OC(=O)\C(=C\C)\C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C34H44O9/c1-9-17(2)31(37)43-25-14-24(41-19(4)35)32(5)16-40-28-29(32)33(25,6)23(13-26(36)38-8)34(7)27-18(3)21(20-10-11-39-15-20)12-22(27)42-30(28)34/h9-11,15,21-25,28-30H,12-14,16H2,1-8H3/b17-9+/t21-,22-,23-,24-,25+,28-,29+,30-,32-,33+,34-/m1/s1
Formula Weight 596.2985
Percent Purity >95%
Chemical Name or Material Salannin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.