missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SALF Antibody (5F12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00286749-M01
This item is not returnable.
View return policy
Description
SALF Monoclonal antibody specifically detects SALF in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| SALF | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| MGC126821, MGC126823, SALF, STON1-GTF2A1L protein, STON1-GTF2A1L readthrough, stoned B/TFIIA-alpha/beta-like factor | |
| SALF (NP_758515, 141 a.a. ∽ 249 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLS | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 5F12 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_758515 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 286749 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction