missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Salmonella typhi hupB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144434
This item is not returnable.
View return policy
Description
Salmonella typhi hupB Polyclonal Antibody for Western Blot, ELISA
Histone-like DNA-binding protein which is capable of wrapping DNA to stabilize it, and thus to prevent its denaturation under extreme environmental conditions.
Specifications
| Salmonella typhi hupB | |
| Polyclonal | |
| Unconjugated | |
| hupB | |
| DNA-binding protein HU-beta; hupB | |
| Rabbit | |
| Protein G | |
| RUO | |
| 57493645, 61281390 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.41 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P0A1R9, P64389 | |
| hupB | |
| Recombinant Neisseria meningitidis serogroup B DNA-binding protein HU-beta(hupB) (1-89aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction