Learn More
Serva Electrophoresis™ Sample Buffer, 2X
Product Code. 11712177
Encompass_Preferred
Change view
Click to view available options
Quantity:
20mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11712177 20mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11712177 Supplier Serva Electrophoresis™ Supplier No. 4253301
 Restricted Item
Estimated Shipment: 11-08-2026
Specifications
Color Blue
Concentration 2X
pH Range 7
Form Solution
For Use With (Application) Native Gel Electrophoresis
Material 1M 6-aminocaproic acid, 100mM BisTris-HCl (pH 7.0), 100mM NaCl, 20% glycerol, 0.1% Serva Blue G
Odor Odorless
Quantity 20mL