missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sarcalumenin Antibody (XIIC4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00€
Specifications
| Antigen | Sarcalumenin |
|---|---|
| Clone | XIIC4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Immunofluorescence 1:500 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18588930
|
Novus Biologicals
NB120-2874-0.025ml |
0.025 mL | |||||||
Description
Sarcalumenin Monoclonal specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunofluorescence.Specifications
| Sarcalumenin | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| sarcalumenin | |
| SRL | |
| IgG1 | |
| Protein A purified | |
| This specifically detects sarcalumenin and the 53 kDa glycoprotein splice variant. |
| XIIC4 | |
| Western Blot 1:500, Immunofluorescence 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Mouse | |
| 6345 | |
| Purified rabbit sarcoplasmic reticulum membrane vesicles. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title