missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine Methyl Ester Hydrochloride, TRC

Product Code. 30412560
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30412560 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30412560

Brand: Toronto Research Chemicals TRCS14052310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sarcosine Methyl Ester Hydrochloride
CAS 13515-93-0
Molecular Formula C4H9NO2 . HCl
Quantity 10 g
InChI Formula InChI=1S/C4H9NO2.ClH/c1-5-3-4(6)7-2;/h5H,3H2,1-2H3;1H
SMILES Cl.CNCC(=O)OC
IUPAC Name methyl 2-(methylamino)acetate;hydrochloride
Molecular Weight (g/mol) 139.58
Formula Weight 139.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.