missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (84C4A), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
558.00€ - 1312.00€
Specifications
| Antigen | SARS-CoV-2 Nucleocapsid |
|---|---|
| Clone | 84C4A |
| Concentration | 0.5 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17117048
|
Invitrogen™
MA537455 |
20 μL |
558.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17136798
![]() |
Invitrogen™
MA537207 |
200 μL |
1312.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.Specifications
| SARS-CoV-2 Nucleocapsid | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.02% sodium azide | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
| N | |
| IgG κ | |
| Protein A | |
| Antibody |
| 84C4A | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| P0DTC9 | |
| 43740575 | |
| SARS-CoV-2 nucleoprotein (NP) specific peptide. | |
| Primary | |
| 4°C | |
| N |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title